3-Chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde 97%

Warning

B23889-03 B23889-06 B23889-06 B23889-03
AAB23889-03EA 45.57 USD
CAAAB23889-03 CAAAB23889-06 AAB23889-03 AAB23889-06
3-Chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde 97%
3-Chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₃ClF₄O
MW: 226.56 g/mol
Boiling Pt: 196 °C
Density: 1.540
Flash Pt: 96 °C(204 °F)
Storage Temperature: Ambient
MDL Number: MFCD01631510
CAS Number: 261763-02-4
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR