Cyclohexanecarboxaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

B23837-14 B23837-14
AAB23837-14EA 75.02 USD
CAAAB23837-14 AAB23837-14
Cyclohexanecarboxaldehyde 97%
Cyclohexanecarboxaldehyde
Formula: C₇H₁₂O
Boiling Pt: 161…163 °C
Melting Pt: 34…35 °C
Density: 0.930
Flash Pt: 40 °C(105 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001457
CAS Number: 2043-61-0
EINECS: 218-057-7
UN: 1989
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR