3-Aminopyrazine-2-carboxylic acid 98+%

Supplier: Thermo Scientific Chemicals

Danger

B23707-06 B23707-06 B23707-14 B23707-03 B23707-14
AAB23707-06EA 60.8 USD
AAB23707-06 CAAAB23707-14 CAAAB23707-06 AAB23707-14 AAB23707-03
3-Aminopyrazine-2-carboxylic acid 98+%
3-Aminopyrazine-2-carboxylic acid
Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
Melting Pt: 204…206 °C (dec.)
Storage Temperature: Ambient
EINECS: 226-558-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR