3-Ethoxypropylamine 99%

Danger

B23694-36 B23694-36
AAB23694-36EA 181.28 USD
AAB23694-36 CAAAB23694-36
3-Ethoxypropylamine 99%
3-Ethoxypropylamine
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 136…138 °C
Density: 0.860
Flash Pt: 32 °C(91 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008221
CAS Number: 6291-85-6
EINECS: 228-545-1
UN: 2920
ADR: "8",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR