Iminodiacetonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning

B23660-22 B23660-36 B23660-0E B23660-0E B23660-22 B23660-36
AAB23660-22EA 35.81 USD
CAAAB23660-0E AAB23660-22 CAAAB23660-36 AAB23660-36 AAB23660-0E CAAAB23660-22
Iminodiacetonitrile 95%
Iminodiacetonitrile
Formula: C₄H₅N₃
MW: 95.11 g/mol
Melting Pt: 75…79 °C
Storage Temperature: Ambient
EINECS: 211-058-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR