Ethyl-3-methyl-3-phenylglycidate (mixture of cis and trans isomers) 98%

Supplier: Thermo Scientific Chemicals

B23654-18 B23654-18 B23654-30 B23654-30 B23654-0B B23654-0B
AAB23654-0BEA 198.69 USD
AAB23654-0B AAB23654-30 CAAAB23654-18 CAAAB23654-30 CAAAB23654-0B AAB23654-18
Ethyl-3-methyl-3-phenylglycidate (mixture of cis and trans isomers) 98%
Ethyl 3-methyl-3-phenylglycidate (mixture of cis and trans isomers)
Formula: C₁₂H₁₄O₃
Boiling Pt: 272…275 °C
Density: 1.044
Flash Pt: 134 °C(273 °F)
Storage Temperature: Ambient
MDL Number: MFCD00022338
CAS Number: 77-83-8
EINECS: 201-061-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR