5-Chloro-2-(trifluoromethyl)phenylacetonitrile 97%

Danger

B23629-06 B23629-03 B23629-06 B23629-03
AAB23629-03EA 72.51 USD
CAAAB23629-06 CAAAB23629-03 AAB23629-03 AAB23629-06
5-Chloro-2-(trifluoromethyl)phenylacetonitrile 97%
5-Chloro-2-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅ClF₃N
Melting Pt: 61…63 °C
Storage Temperature: Ambient
MDL Number: MFCD01631357
CAS Number: 261763-26-2
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR