3-Chloro-4-[(trifluoromethyl)thio]aniline 97%

Supplier: Thermo Scientific Chemicals

Danger

B23500-03 B23500-03 B23500-06 B23500-06
AAAB23500-03EA 83.3 USD
CAAAAB23500-03 CAAAAB23500-06 AAAB23500-03 AAAB23500-06
3-Chloro-4-[(trifluoromethyl)thio]aniline 97%
3-Chloro-4-[(trifluoromethyl)thio]aniline
Formula: C₇H₅ClF₃NS
Melting Pt: 41…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00190131
CAS Number: 64628-74-6
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR