3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) 97%

Danger

Synonyms: 3,5-Dimethylpiperidine

B23435-14 B23435-22 B23435-14
AAAB23435-14EA 51.96 USD
CAAAAB23435-22 CAAAAB23435-14 AAAB23435-14
3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) 97%
3,5-Dimethylpiperidin (mixture of cis- and trans-isomers)
Formula: C₇H₁₅N
MW: 113.20 g/mol
Boiling Pt: 142…146 °C
Density: 0.830
Flash Pt: 32 °C(89 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005996
CAS Number: 35794-11-7
EINECS: 252-730-6
UN: 2733
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR