Methyl-2-benzyloxybenzoate 98%

B23309-06 B23309-22 B23309-14 B23309-06 B23309-22 B23309-14
AAB23309-06EA 38.7 USD
AAB23309-06 CAAAB23309-22 CAAAB23309-14 CAAAB23309-06 AAB23309-14 AAB23309-22
Methyl-2-benzyloxybenzoate 98%
Methyl 2-benzyloxybenzoate
Formula: C₁₅H₁₄O₃
Melting Pt: 45…48 °C
Storage Temperature: Ambient
MDL Number: MFCD01099045
CAS Number: 55142-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR