2-Chloro-3-(trifluoromethyl)benzyl alcohol 97+%

Warning

Synonyms: (2-Chloro-3-(trifluoromethyl)phenyl)methanol

B23212-03 B23212-06 B23212-03 B23212-06
AAB23212-03EA 65.27 USD
AAB23212-03 CAAAB23212-06 AAB23212-06 CAAAB23212-03
2-Chloro-3-(trifluoromethyl)benzyl alcohol 97+%
2-Chloro-3-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆ClF₃O
MW: 210.58 g/mol
Melting Pt: 74…77 °C
Storage Temperature: Ambient
MDL Number: MFCD01631537
CAS Number: 261763-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR