Benzyl-4-oxo-1-piperidinecarboxylate ≥98%

Warning

Synonyms: 1-Benzyloxycarbonyl-4-piperidine1-Carbobenzoxy-4-piperidone1-Cbz-4-piperidone4-Oxo-1-piperidinecarboxylic Acid Benzyl Ester1-Cbz-Piperidin-4-one

B23181-03 B23181-06 B23181-06 B23181-03 B23181-14
AAB23181-06EA 55.69 USD
AAB23181-06 AAB23181-03 AAB23181-14 CAAAB23181-03 CAAAB23181-06
Benzyl-4-oxo-1-piperidinecarboxylate ≥98%
Benzyl-4-oxo-1-piperidinecarboxylate
Formula: C₁₃H₁₅NO₃
MW: 233.27 g/mol
Melting Pt: 38…41 °C
Density: 1.172
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00673144
CAS Number: 19099-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR