5-Chloro-3-phenyl-2,1-benzisoxazole 98+%

Warning

Synonyms: 5-Chloro-3-phenylbenzo[c]isoxazole

B23130-09 B23130-18 B23130-30 B23130-09 B23130-18 B23130-30
AAB23130-30EA 221.11 USD
CAAAB23130-09 CAAAB23130-18 AAB23130-30 AAB23130-09 CAAAB23130-30 AAB23130-18
5-Chloro-3-phenyl-2,1-benzisoxazole 98+%
5-Chloro-3-phenyl-2,1-benzisoxazole
Formula: C₁₃H₈ClNO
MW: 229.67 g/mol
Melting Pt: 114…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00014573
CAS Number: 719-64-2
EINECS: 211-950-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR