1-Furfurylpyrrole 99%

Warning

Synonyms: 1-(Furan-2-ylmethyl)-1H-pyrrole

B23105-22 B23105-14 B23105-06 B23105-06 B23105-14 B23105-22
AAB23105-14EA 228.79 USD
AAB23105-14 AAB23105-22 CAAAB23105-06 CAAAB23105-22 CAAAB23105-14 AAB23105-06
1-Furfurylpyrrole 99%
1-Furfurylpyrrole
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 76…78 °C/1 mm
Density: 1.081
Flash Pt: 92 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010080
CAS Number: 1438-94-4
EINECS: 215-876-1
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR