2-Phenylbutyryl chloride 98%
|
Danger
|
B22997-22
B22997-14
B22997-14
B22997-22
AAB22997-14EA
94.18
USD
AAB22997-14
CAAAB22997-14
AAAB22997-22
CAAAAB22997-22
2-Phenylbutyryl chloride 98%
2-Phenylbutyryl chloride
Formula:
C₁₀H₁₁ClO MW: 182.65 Boiling Pt: 143…145 °C/15 mm Density: 1.093 Flash Pt: 98 °C(208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00018811 CAS Number: 36854-57-6 EINECS: 253-241-0 UN: 3265 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...