6-Chloro-2-fluoro-3-methylbenzaldehyde 97%

Warning

B22937-06 B22937-03 B22937-06 B22937-03
AAB22937-06EA 128.66 USD
AAB22937-06 CAAAB22937-03 CAAAB22937-06 AAB22937-03
6-Chloro-2-fluoro-3-methylbenzaldehyde 97%
6-Chloro-2-fluoro-3-methylbenzaldehyde
Formula: C₈H₆ClFO
MW: 172.59 g/mol
Boiling Pt: 249 °C
Density: 1.300
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01631370
CAS Number: 286474-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR