2'-Fluoro-3'-(trifluoromethyl)acetophenone 97%

Warning

Synonyms: 1-(2-Fluoro-3-(trifluoromethyl)phenyl)ethanone

B22931-06 B22931-03 B22931-06 B22931-03
AAB22931-06EA 281.26 USD
CAAAB22931-03 AAB22931-06 AAB22931-03 CAAAB22931-06
2'-Fluoro-3'-(trifluoromethyl)acetophenone 97%
2'-Fluoro-3'-(trifluoromethyl)acetophenone
Formula: C₉H₆F₄O
MW: 206.14 g/mol
Density: 1,020
Storage Temperature: Ambient
MDL Number: MFCD00061145
CAS Number: 207853-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR