4'-(1-Piperazinyl)acetophenone 94%

Danger

Synonyms: 4-Piperazinoacetophenone

B22832-06 B22832-22 B22832-22 B22832-14 B22832-06 B22832-14
AAB22832-14EA 192.61 USD
CAAAB22832-22 CAAAB22832-14 AAB22832-14 CAAAB22832-06 AAB22832-22 AAB22832-06
4'-(1-Piperazinyl)acetophenone 94%
4'-(1-Piperazinyl)acetophenone
Formula: C₁₂H₁₆N₂O
MW: 204.27
Melting Pt: 93…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00005962
CAS Number: 51639-48-6
EINECS: 257-332-6
UN: 3259
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR