(Furfurylthio)acetone 96%
|
Warning
|
B22724-14
B22724-14
B22724-06
B22724-06
AAB22724-14EA
200.55
USD
AAB22724-14
CAAAB22724-06
CAAAB22724-14
AAB22724-06
(Furfurylthio)acetone 96%
(Furfurylthio)acetone
Formula:
C₈H₁₀O₂S Storage Temperature: Ambient |
MDL Number:
MFCD02683098 CAS Number: 58066-86-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...