Methyl (2-furfurylthio)acetate 97%

Warning

B22717-06 B22717-06 B22717-22 B22717-22 B22717-14 B22717-14
AAB22717-14EA 95.79 USD
CAAAB22717-06 AAB22717-14 AAB22717-22 AAB22717-06 CAAAB22717-14 CAAAB22717-22
Methyl (2-furfurylthio)acetate 97%
Methyl (2-furfurylthio)acetate
Formula: C₈H₁₀O₂S
Boiling Pt: 95 °C/3 mm
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD02283460
CAS Number: 108499-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR