2,4-Dimethyl-2-pentenoic acid, predominantly trans 99%

Danger

B22701-88 B22701-88 B22701-09 B22701-09
AAB22701-09EA 211.09 USD
CAAAB22701-88 AAB22701-09 AAB22701-88 CAAAB22701-09
2,4-Dimethyl-2-pentenoic acid, predominantly trans 99%
2,4-Dimethyl-2-pentenoic acid, predominantly trans
Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Density: 0.949
Storage Temperature: Ambient
MDL Number: MFCD00209505
CAS Number: 66634-97-7
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR