2,5-Dimethoxybenzenesulfonamide 97%

B22649-03 B22649-06 B22649-03 B22649-06
AAB22649-06EA 378.51 USD
AAB22649-06 CAAAB22649-03 AAB22649-03 CAAAB22649-06
2,5-Dimethoxybenzenesulfonamide 97%
2,5-Dimethoxybenzenesulphonamide
Formula: C₈H₁₁NO₄S
Melting Pt: 145…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00833415
CAS Number: 19116-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR