1-(Methylamino)-1-(methylthio)-2-nitroethene 97%

Warning

Synonyms: 1-Methylthio-1-methylamino-2-nitroethyleneN-Methyl-1-(methylthio)-2-nitroethenamine

B22608-09 B22608-18 B22608-30 B22608-18 B22608-30 B22608-09
AAB22608-09EA 23.69 USD
CAAAB22608-30 AAB22608-09 AAB22608-18 CAAAB22608-09 AAB22608-30 CAAAB22608-18
1-(Methylamino)-1-(methylthio)-2-nitroethene 97%
1-(Methylamino)-1-(methylthio)-2-nitroethene
Formula: C₄H₈N₂O₂S
MW: 148.18 g/mol
Melting Pt: 109…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00040994
CAS Number: 61832-41-5
EINECS: 263-266-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR