9-Dodecyn-1-ol 97%

B22577-06 B22577-14 B22577-14 B22577-06
AAB22577-14EA 448.87 USD
AAB22577-14 CAAAB22577-06 CAAAB22577-14 AAB22577-06
9-Dodecyn-1-ol 97%
9-Dodecyn-1-ol
Formula: C₁₂H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD00082940
CAS Number: 71084-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR