4,5-Dimethyl-2-isobutylthiazole 99%

Warning

B22554-88 B22554-09 B22554-88 B22554-09
AAB22554-09EA 56.76 USD
CAAAB22554-09 AAB22554-09 AAB22554-88 CAAAB22554-88
4,5-Dimethyl-2-isobutylthiazole 99%
4,5-Dimethyl-2-isobutylthiazole
Formula: C₉H₁₅NS
MW: 169.28
Density: 0,967
Storage Temperature: Ambient
MDL Number: MFCD00053136
CAS Number: 53498-32-1
EINECS: 258-586-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR