5-Cyano-1H-imidazole-4-carboxamide hydrate 97%

Warning

B22535-03 B22535-06 B22535-03 B22535-06
AAB22535-06EA 320.79 USD
CAAAB22535-03 AAB22535-06 CAAAB22535-06 AAB22535-03
5-Cyano-1H-imidazole-4-carboxamide hydrate 97%
5-Cyano-1H-imidazole-4-carboxamide hydrate
Formula: C₅H₄N₄O
MW: 136.11 g/mol
Melting Pt: ca 272 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD04038014
CAS Number: 5372-23-6
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR