Methyl-4-amino-3-chlorobenzoate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 4-amino-3-chlorobenzoate

B22519-03 B22519-06 B22519-03 B22519-06
AAB22519-06EA 166.99 USD
AAB22519-06 CAAAB22519-03 AAB22519-03 CAAAB22519-06
Methyl-4-amino-3-chlorobenzoate 99%
Methyl 4-amino-3-chlorobenzoate
Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Melting Pt: 112…113 °C
Storage Temperature: Ambient
MDL Number: MFCD01320686
CAS Number: 84228-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR