1-Acetylpyrene 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(Pyren-1-yl)ethanone

B22483-88 B22483-88
AAB22483-88EA 62.39 USD
AAB22483-88 CAAAB22483-88
1-Acetylpyrene 97%
1-Acetylpyrene
Formula: C₁₈H₁₂O
MW: 244.29 g/mol
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00021663
CAS Number: 3264-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR