Methyl-4-(chloromethyl)benzoate 97%

Supplier: Thermo Scientific Chemicals

Danger

Synonyms: Methyl 4-(chloromethyl)benzoate

B22476-09 B22476-04 B22476-04 B22476-09
AAB22476-04EA 56.6 USD
CAAAB22476-04 CAAAB22476-09 AAB22476-04 AAB22476-09
Methyl-4-(chloromethyl)benzoate 97%
Methyl 4-(chloromethyl)benzoate
Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Boiling Pt: 160…165 °C/30 mm
Melting Pt: 39…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00511224
CAS Number: 34040-64-7
EINECS: 251-806-6
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR