2-Carboxyphenyl salicylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Salicyloyloxybenzoic AcidO-Salicyloylsalicylic AcidSalicylic Acid 2-Carboxyphenyl EsterSalsalate

B22444-14 B22444-36 B22444-22 B22444-22 B22444-36 B22444-14
AAB22444-14EA 64.69 USD
AAB22444-14 AAB22444-36 AAB22444-22 CAAAB22444-36 CAAAB22444-14 CAAAB22444-22
2-Carboxyphenyl salicylate 98%
2-Carboxyphenyl salicylate
Formula: C₁₄H₁₀O₅
MW: 258.23 g/mol
Melting Pt: 132…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00020252
CAS Number: 552-94-3
EINECS: 209-027-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR