1,3-Bis(methylthio)propane 97%

B22397-03 B22397-06
AAAB22397-03EA 88.9 USD
AAAB22397-03 AAAB22397-06
1,3-Bis(methylthio)propane 97%
1,3-Bis(methylthio)propane
Formula: C₅H₁₂S₂
Boiling Pt: 88…90 °C/12 mm
Density: 1,011
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039811
CAS Number: 24949-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR