3-Nitro-4-(1-piperidinyl)benzaldehyde 96%

Warning

B22381-03 B22381-06 B22381-03 B22381-06
AAB22381-03EA 49.61 USD
AAB22381-03 AAB22381-06 CAAAB22381-06 CAAAB22381-03
3-Nitro-4-(1-piperidinyl)benzaldehyde 96%
3-Nitro-4-(1-piperidinyl)benzaldehyde
Formula: C₁₂H₁₄N₂O₃
MW: 234.26
Melting Pt: 50…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00831780
CAS Number: 39911-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR