2-Diphenylphosphino-1-naphthoic acid 97%

Warning

B22361-14 B22361-06 B22361-03 B22361-03 B22361-06 B22361-14
AAB22361-14EA 1300.66 USD
AAB22361-14 AAB22361-03 AAB22361-06 CAAAB22361-06 CAAAB22361-03 CAAAB22361-14
2-Diphenylphosphino-1-naphthoic acid 97%
2-Diphenylphosphino-1-naphthoic acid
Formula: C₂₃H₁₇O₂P
MW: 356.36 g/mol
Melting Pt: 190…192 °C
Storage Temperature: Ambient
MDL Number: MFCD02683090
CAS Number: 178176-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR