2-Methyl-5-nitro-2H-indazole 98+%

B22360-01 B22360-06 B22360-03 B22360-01 B22360-06 B22360-03
AAB22360-03EA 109.78 USD
AAB22360-03 CAAAAB22360-06 AAB22360-01 AAAB22360-06 CAAAB22360-03 CAAAB22360-01
2-Methyl-5-nitro-2H-indazole 98+%
2-Methyl-5-nitro-2H-indazole
Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
Melting Pt: 126…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00051809
CAS Number: 5228-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR