4-Cyano-4'-ethylbiphenyl 99%
|
Warning
|
Synonyms:
4'-Ethyl-[1,1'-biphenyl]-4-carbonitrile
B22359-06
B22359-06
B22359-03
B22359-03
AAB22359-03EA
54.6
USD
CAAAB22359-06
AAB22359-03
AAB22359-06
CAAAB22359-03
4-Cyano-4'-ethylbiphenyl 99%
4-Cyano-4'-ethylbiphenyl
Formula:
C₁₅H₁₃N MW: 207.27 g/mol Boiling Pt: 202…205 °C/3 mm Melting Pt: 74…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00799420 CAS Number: 58743-75-2 EINECS: 261-414-7 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...