(Triphenylmethyl)triphenylphosphonium chloride 98%

Warning

B22319-09 B22319-18 B22319-18 B22319-09 B22319-30 B22319-30
AAB22319-30EA 683.76 USD
AAB22319-30 CAAAB22319-18 CAAAB22319-09 AAB22319-18 CAAAB22319-30 AAB22319-09
(Triphenylmethyl)triphenylphosphonium chloride 98%
(Triphenylmethyl)triphenylphosphonium chloride
Formula: C₃₇H₃₀ClP
Melting Pt: 244…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00082244
CAS Number: 48234-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR