4-Butylbenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (4-Butylphenyl)methanamine

B22316-22 B22316-22 B22316-14 B22316-14
AAB22316-22EA 1888.02 USD
AAB22316-22 CAAAB22316-14 AAB22316-14 CAAAB22316-22
4-Butylbenzylamine 98%
4-Butylbenzylamine
Formula: C₁₁H₁₇N
MW: 163.26 g/mol
Boiling Pt: 89…91 °C/1 mm
Storage Temperature: Ambient
MDL Number: MFCD02258855
CAS Number: 57802-79-6
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR