2,4,5-Trimethyl-3-oxazoline cis- and trans- mixture 99%

Warning

B22228-06 B22228-06 B22228-14 B22228-14 B22228-22 B22228-22
AAB22228-14EA 101.56 USD
AAB22228-14 AAB22228-06 CAAAB22228-22 CAAAB22228-14 AAB22228-22 CAAAB22228-06
2,4,5-Trimethyl-3-oxazoline cis- and trans- mixture 99%
2,4,5-Trimethyl-3-oxazoline
Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 45…47 °C/12 mm
Storage Temperature: Ambient
MDL Number: MFCD00053161
CAS Number: 22694-96-8
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR