Benzyl-4-bromophenyl ketone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(4-Bromophenyl)-2-phenylethanone
B22159-06
B22159-06
B22159-03
B22159-03
AAB22159-06EA
189.99
USD
AAB22159-06
CAAAB22159-06
CAAAB22159-03
AAB22159-03
Benzyl-4-bromophenyl ketone 98%
Benzyl 4-bromophenyl ketone
Formula:
C₁₄H₁₁BrO MW: 275.14 g/mol Melting Pt: 109…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016331 CAS Number: 2001-29-8 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...