1-(Dimethylcarbamoyl)-4-(2-sulfoethyl)pyridinium hydroxide inner salt 95%

Warning

Synonyms: 2-[1-(Dimethylcarbamoyl)-4-pyridinio]ethylsulfonate

B22124-09 B22124-30 B22124-18 B22124-18 B22124-30 B22124-09
AAB22124-09EA 24.63 USD
AAB22124-09 CAAAB22124-30 AAB22124-18 AAB22124-30 CAAAB22124-09 CAAAB22124-18
1-(Dimethylcarbamoyl)-4-(2-sulfoethyl)pyridinium hydroxide inner salt 95%
1-(Dimethylcarbamoyl)-4-(2-sulfoethyl)pyridinium hydroxide inner salt
Formula: C₁₀H₁₄N₂O₄S
MW: 258.29 g/mol
Melting Pt: ca 180 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00185917
CAS Number: 136997-71-2
EINECS: 418-440-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR