1-Benzoylnaphthalene 97%

Warning

Synonyms: 1-Naphthyl phenyl ketone

B22120-03 B22120-06 B22120-06 B22120-03 B22120-14 B22120-14
AAB22120-03EA 191.73 USD
CAAAB22120-06 AAB22120-03 AAB22120-14 AAB22120-06 CAAAB22120-03 CAAAB22120-14
1-Benzoylnaphthalene 97%
1-Benzoylnaphthalene
Formula: C₁₇H₁₂O
MW: 232.28
Melting Pt: 75…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00046431
CAS Number: 642-29-5
EINECS: 211-382-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR