4-Hydroxy-4'-iodobiphenyl 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22115-22
B22115-06
B22115-14
B22115-14
B22115-22
B22115-06
AAB22115-06EA
69.59
USD
CAAAB22115-22
CAAAB22115-14
AAB22115-06
AAB22115-14
AAB22115-22
CAAAB22115-06
4-Hydroxy-4'-iodobiphenyl 98+%
4-Hydroxy-4'-iodobiphenyl
Formula:
C₁₂H₉IO Melting Pt: 196…202°C Storage Temperature: Ambient |
MDL Number:
MFCD02258843 CAS Number: 29558-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...