4-Methyl-4-(methylthio)-2-pentanone 99%

Warning

Synonyms: 4-Methyl-4-methylthio-2-pentanone4-Methyl-4-(methylthio)pentan-2-one

B22058-14 B22058-22 B22058-22 B22058-14
AAB22058-14EA 65.68 USD
CAAAB22058-14 AAB22058-14 AAB22058-22 CAAAB22058-22
4-Methyl-4-(methylthio)-2-pentanone 99%
4-Methyl-4-(methylthio)-2-pentanone
Formula: C₇H₁₄OS
MW: 146.25 g/mol
Boiling Pt: 80…82 °C/10 mm
Density: 0.965
Flash Pt: 72 °C(162 °F)
Storage Temperature: Ambient
MDL Number: MFCD00049253
CAS Number: 23550-40-5
EINECS: 245-727-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR