3-Chloro-4-methylphenylhydrazine hydrochloride 97%

Warning

Synonyms: 3-Chloro-4-methylphenylhydrazine Hydrochloride

B22053-03 B22053-06 B22053-03 B22053-06
AAB22053-03EA 38.63 USD
CAAAB22053-06 CAAAB22053-03 AAB22053-03 AAB22053-06
3-Chloro-4-methylphenylhydrazine hydrochloride 97%
3-Chloro-4-methylphenylhydrazine hydrochloride
Formula: C₇H₉ClN₂.HCl
MW: 193.08 g/mol
Melting Pt: ca 233 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012936
CAS Number: 54812-56-5
EINECS: 259-357-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR