4-Cyano-4'-heptyloxybiphenyl 99%
Synonyms:
4'-(Heptyloxy)-[1,1'-biphenyl]-4-carbonitrile
B22044-03
B22044-06
B22044-03
B22044-06
AAB22044-03EA
67.11
USD
CAAAB22044-03
CAAAB22044-06
AAB22044-03
AAB22044-06
4-Cyano-4'-heptyloxybiphenyl 99%
4-Cyano-4'-heptyloxybiphenyl
Formula:
C₂₀H₂₃NO MW: 293.41 g/mol Melting Pt: 53…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134146 CAS Number: 52364-72-4 EINECS: 257-876-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...