Trimethylsilyl acetate (acetoxytrimethylsilane) 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylsilyl acetate

B22037-22 B22037-22
AAB22037-22EA 88.79 USD
AAB22037-22 CAAAB22037-22
Trimethylsilyl acetate (acetoxytrimethylsilane) 97%
Trimethylsilyl acetate (Acetoxytrimethylsilane)
Formula: C₅H₁₂O₂Si
MW: 132.24 g/mol
Boiling Pt: 107 °C
Melting Pt: –32 °C
Density: 0.884
Flash Pt: 4 °C(39 °F)
Storage Temperature: Ambient
EINECS: 220-404-2
UN: 3272
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR