4'-Bromo-3'-methylacetanilide 98%

B21980-22 B21980-14 B21980-14 B21980-06 B21980-22 B21980-06
AAB21980-06EA 25.99 USD
CAAAB21980-22 AAB21980-06 AAB21980-14 CAAAB21980-06 AAB21980-22 CAAAB21980-14
4'-Bromo-3'-methylacetanilide 98%
4'-Bromo-3'-methylacetanilide
Formula: C₉H₁₀BrNO
Melting Pt: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00040822
CAS Number: 90914-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR