Ethyl-2,3,4,5-tetrafluorobenzoate 98%

Synonyms: Ethyl 2,3,4,5-tetrafluorobenzoate

B21920-04 B21920-04 B21920-09 B21920-09
AAB21920-09EA 146.52 USD
AAB21920-09 CAAAB21920-04 AAB21920-04 CAAAB21920-09
Ethyl-2,3,4,5-tetrafluorobenzoate 98%
Ethyl 2,3,4,5-tetrafluorobenzoate
Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02094158
CAS Number: 122894-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR