2-Chloro-5-n-decylpyrimidine 99%

Warning

B21889-03 B21889-06 B21889-06 B21889-03
AAB21889-06EA 314.13 USD
CAAAB21889-06 CAAAB21889-03 AAB21889-06 AAB21889-03
2-Chloro-5-n-decylpyrimidine 99%
2-Chloro-5-n-decylpyrimidine
Formula: C₁₄H₂₃ClN₂
MW: 254.8 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683077
CAS Number: 170434-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR