(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid 96%
|
Warning
|
Synonyms:
[(5-Sulphanyl-1,3,4-thiadiazol-2-yl)sulphanyl]acetic acid
B21882-06
B21882-03
B21882-06
B21882-03
AAB21882-06EA
182.04
USD
AAB21882-06
CAAAB21882-03
AAB21882-03
CAAAB21882-06
(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid 96%
(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid
Formula:
C₄H₄N₂O₂S₃ MW: 208.28 g/mol Melting Pt: 165…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00129970 CAS Number: 53723-88-9 EINECS: 258-728-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...